Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006938.3 | NP_008869.1 | 3 | 353 | ||
GenBank mRNA | AB451227.1 | - | 1 | 192 | ||
AB451350.1 | - | 1 | 192 | |||
AJ577268.1 | - | 3 | 285 | |||
BC001721.1 | - | 3 | 314 | 3355716 | ||
BC072427.1 | - | 3 | 240 | 6464946 | ||
BU565530.1 | - | 3 | 355 | |||
CR542239.1 | - | 1 | 192 | |||
HY043971.1 | - | 3 | 351 | |||
J03798.1 | - | 3 | 342 |