Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pnliprp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032081.1 | NP_114470.1 | 5 | 462 | ||
GenBank mRNA | AB072249.1 | - | 3 | 235 | ||
FQ232772.1 | - | 5 | 461 | |||
X61925.1 | - | 5 | 462 |