Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053587.1 | NP_446039.1 | 2 | 423 | ||
XM_006232780.1 | XP_006232842.1 | 3 | 505 | |||
GenBank mRNA | L18948.1 | - | 2 | 423 |