Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BTN3A2 SIRNA Assays ›
Gene Symbol:
BTN3A2
Gene Name:
butyrophilin subfamily 3 member A2
Gene Aliases:
BT3.2, BTF4, BTN3.2, CD277
Chromosome Location:
Chr.6: 26365159 - 26451697 on Build GRCh38
UniGene ID:
Hs.376046
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
19034
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BTN3A2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001197246.2
NP_001184175.1
5, 6
940
NM_001197247.2
NP_001184176.1
7, 8
1205
NM_001197248.2
NP_001184177.1
5, 6
1044
NM_001197249.2
NP_001184178.1
6, 7
1105
NM_007047.4
NP_008978.2
7, 8
1195
XM_005248827.3
XP_005248884.1
7, 8
1183
XM_005248831.3
XP_005248888.1
6, 7
982
XM_005248832.3
XP_005248889.1
6, 7
982
XM_006714979.3
XP_006715042.1
7, 8
1198
XM_006714980.3
XP_006715043.1
7, 8
1198
XM_006714981.3
XP_006715044.1
7, 8
1188
XM_006714982.3
XP_006715045.1
6, 7
1108
XM_011514267.2
XP_011512569.1
7, 8
1064
XM_011514268.2
XP_011512570.1
7, 8
1198
XM_011514269.2
XP_011512571.1
6, 7
967
XM_011514270.2
XP_011512572.1
6, 7
901
XM_011514271.2
XP_011512573.1
6, 7
767
XM_017010211.1
XP_016865700.1
7, 8
1173
XM_017010212.1
XP_016865701.1
7, 8
1049
XM_017010213.1
XP_016865702.1
6, 7
1070
XM_017010214.1
XP_016865703.1
6, 7
1093
XM_017010215.1
XP_016865704.1
6, 7
901
XM_017010216.1
XP_016865705.1
5, 6
892
XM_017010217.1
XP_016865706.1
5, 6
811
GenBank mRNA
AF257505.1
-
5, 6
942
AK290880.1
-
7, 8
1133
AK299417.1
-
6, 7
989
AK303600.1
-
5, 6
961
AK312847.1
-
7, 8
1127
BC002832.2
-
7, 8
1086
3637548
BC020214.1
-
7, 8
1086
4870774
U90144.1
-
6, 7
1002
U90546.1
-
7, 8
1123
Back To Top
More Information
Panther Classification:
Molecular Function -
immunoglobulin receptor superfamily
Gene Ontology Categories:
Function(s)
T cell mediated immunity
interferon-gamma secretion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-7q954:80/10.239.162.42:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0