Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P3h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053667.1 | NP_446119.1 | 2 | 493 | ||
GenBank mRNA | AF087433.1 | - | 2 | 493 | ||
BC161838.1 | - | 2 | 527 | 9101602 |
Gene Symbol: P3h3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006237343.2 | XP_006237405.1 | 2 | 608 | ||
XM_006237344.2 | XP_006237406.1 | 2 | 623 |