Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053670.3 | NP_446122.1 | 6 | 598 | ||
GenBank mRNA | AF440799.2 | - | 6 | 598 | ||
BC059117.1 | - | 6 | 702 | 6891094 |
Set Membership: |
G-protein coupled receptor |