Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053681.1 | NP_446133.1 | 3 | 315 | ||
XM_006232558.2 | XP_006232620.1 | 3 | 518 | |||
XM_006232559.2 | XP_006232621.1 | 3 | 1272 | |||
XM_006232560.2 | XP_006232622.1 | 3 | 1721 | |||
XM_006232563.2 | XP_006232625.1 | 3 | 303 | |||
XM_008761122.1 | XP_008759344.1 | 4 | 750 | |||
XM_008761123.1 | XP_008759345.1 | 3 | 793 | |||
GenBank mRNA | AF140231.1 | - | 3 | 315 |