Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erp29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053961.2 | NP_446413.1 | 2 | 230 | ||
GenBank mRNA | BC091129.1 | - | 2 | 230 | 7308437 | |
FQ213671.1 | - | 2 | 228 | |||
FQ218939.1 | - | 2 | 228 | |||
FQ233694.1 | - | 2 | 228 | |||
FQ234786.1 | - | 2 | 228 | |||
U36482.1 | - | 2 | 258 | |||
Y10264.1 | - | 2 | 229 |