Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Agpat4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133406.1 | NP_596897.1 | 2, 3 | 391 | ||
XM_006227856.2 | XP_006227918.1 | 2, 3 | 1106 | |||
XM_006227857.2 | XP_006227919.1 | 2, 3 | 760 | |||
GenBank mRNA | AB067572.1 | - | 2, 3 | 391 | ||
BC086992.1 | - | 2, 3 | 310 | 7191675 |