Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcer2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033924.1 | NP_001029096.1 | 4 | 274 | ||
NM_133550.2 | NP_598234.2 | 4 | 274 | |||
XM_008768939.1 | XP_008767161.1 | 3 | 438 | |||
GenBank mRNA | AF381978.1 | - | 3 | 160 | ||
BC078749.1 | - | 4 | 274 | 7124300 | ||
X73579.1 | - | 3 | 157 |