Search Thermo Fisher Scientific
Gene Symbol: Ero1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138528.1 | NP_612537.1 | 5 | 396 | ||
GenBank mRNA | AF489855.1 | - | 5 | 383 | ||
AY071924.1 | - | 5 | 396 | |||
AY071925.1 | - | 2 | 78 | |||
FQ228432.1 | - | 241 |