Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138849.1 | NP_620204.1 | 8 | 1535 | ||
XM_006230101.2 | XP_006230163.1 | 8 | 1630 | |||
XM_006230102.2 | XP_006230164.1 | 8 | 1515 | |||
XM_006230104.1 | XP_006230166.1 | 8 | 1729 | |||
XM_008759675.1 | XP_008757897.1 | 6 | 1995 | |||
GenBank mRNA | U30831.1 | - | 8 | 1535 |