Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270762.1 | NP_001257691.1 | 5 | 619 | ||
NM_001270763.1 | NP_001257692.1 | 5 | 625 | |||
NM_139105.3 | NP_620805.2 | 5 | 634 | |||
XM_008759970.1 | XP_008758192.1 | 5 | 1183 | |||
GenBank mRNA | AB296102.1 | - | 4 | 501 | ||
BC070501.1 | - | 5 | 541 | 7096144 | ||
FQ233708.1 | - | 5 | 559 | |||
FQ234889.1 | - | 5 | 558 | |||
X62528.1 | - | 5 | 619 |