Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007030.2 | NP_008961.1 | 4 | 1430 | ||
XM_005248237.3 | XP_005248294.2 | 4 | 1495 | |||
XM_017008993.1 | XP_016864482.1 | 4 | 3626 | |||
XM_017008994.1 | XP_016864483.1 | 6 | 1481 | |||
GenBank mRNA | AB017016.1 | - | 4 | 1341 | ||
AK054920.1 | - | 3 | 2027 | |||
AK098392.1 | - | 4 | 1386 | |||
BC040496.1 | - | 4 | 1354 | |||
BC131506.1 | - | 3 | 1326 | 40082752 |