Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145677.1 | NP_663710.1 | 3, 4 | 482 | ||
XM_006233932.2 | XP_006233994.1 | 3, 4 | 603 | |||
XM_006233934.2 | XP_006233996.1 | 3, 4 | 1255 | |||
GenBank mRNA | AY043169.1 | - | 3, 4 | 482 |