Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Miox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145771.2 | NP_665714.2 | 4, 5 | 353 | ||
XM_008765674.1 | XP_008763896.1 | 4, 5 | 537 | |||
GenBank mRNA | AF197128.1 | - | 4, 5 | 347 | ||
AF230096.1 | - | 3, 4 | 355 | |||
AY738259.1 | - | 3, 4 | 347 | |||
BC078840.1 | - | 4, 5 | 353 | 7110224 | ||
FQ219187.1 | - | 3, 4 | 342 |