Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_171990.2 | NP_741987.1 | 7, 8 | 742 | ||
XM_006250429.2 | XP_006250491.1 | 8, 9 | 829 | |||
XM_006250430.2 | XP_006250492.1 | 8, 9 | 884 | |||
XM_006250431.2 | XP_006250493.1 | 7, 8 | 838 | |||
GenBank mRNA | AJ000347.1 | - | 7, 8 | 693 | ||
BC085692.1 | - | 7, 8 | 742 | 7190529 | ||
FQ214355.1 | - | 8, 9 | 810 |