Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naglt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176080.2 | NP_788269.1 | 2 | 328 | ||
GenBank mRNA | AB089802.1 | - | 2 | 328 | ||
BC098651.1 | - | 2 | 315 | 7130354 |
Gene Symbol: RGD1561777 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773019.1 | XP_008771241.1 | 161 |