Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008396.2 | NP_001008397.1 | 2 | 173 | ||
NM_007086.3 | NP_009017.1 | 3 | 266 | |||
XM_006720012.2 | XP_006720075.1 | 3 | 264 | |||
XM_011536373.2 | XP_011534675.1 | 2 | 177 | |||
XM_017020940.1 | XP_016876429.1 | 2 | 177 | |||
GenBank mRNA | AJ006266.1 | - | 2 | 215 | ||
AK293005.1 | - | 3 | 233 | |||
BC000622.2 | - | 3 | 231 | |||
BC005040.1 | - | 3 | 231 | |||
BC043349.1 | - | 3 | 254 | |||
BC063041.1 | - | 3 | 242 | 6514641 | ||
DA189308.1 | - | 2 | 173 | |||
DB082418.1 | - | 3 | 266 |