Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001834.3 | NP_001825.1 | 5 | 687 | ||
NM_007097.3 | NP_009028.1 | 6 | 741 | |||
NR_045724.1 | 6 | 1239 | ||||
XM_017009026.1 | XP_016864515.1 | 5 | 467 | |||
GenBank mRNA | AK300200.1 | - | 6 | 1209 | ||
BC006332.2 | - | 5 | 625 | 4299637 | ||
BC006457.1 | - | 5 | 611 | 3538261 | ||
BT007028.1 | - | 5 | 482 | |||
CR536577.1 | - | 5 | 482 | |||
M20469.1 | - | 6 | 708 | |||
M20470.1 | - | 5 | 621 |