Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp191 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182955.1 | NP_892000.1 | 2 | 269 | ||
XM_006254464.2 | XP_006254526.1 | 2 | 306 | |||
XM_006254465.2 | XP_006254527.1 | 2 | 664 | |||
GenBank mRNA | AY336973.1 | - | 2 | 269 |