Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Art2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198735.2 | NP_942030.1 | 2 | 255 | ||
XM_008759694.1 | XP_008757916.1 | 2 | 483 | |||
XM_008759695.1 | XP_008757917.1 | 2 | 483 | |||
XM_008759696.1 | XP_008757918.1 | 2 | 1104 | |||
XM_008759697.1 | XP_008757919.1 | 3 | 1472 | |||
XM_008759698.1 | XP_008757920.1 | 5 | 844 | |||
XM_008759699.1 | XP_008757921.1 | 2 | 330 | |||
XM_008759700.1 | XP_008757922.1 | 4 | 575 | |||
XM_008759701.1 | XP_008757923.1 | 2 | 398 | |||
XM_008759702.1 | XP_008757924.1 | 2 | 1104 | |||
GenBank mRNA | BC099070.1 | - | 2 | 255 | 7366134 | |
FQ227811.1 | - | 1 | 28 | |||
M85193.1 | - | 2 | 232 | |||
X52082.1 | - | 2 | 225 | |||
X99122.1 | - | 5 | 849 | |||
X99123.1 | - | 3 | 305 | |||
X99124.1 | - | 3 | 322 |