Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MYL1 SIRNA Assays ›
Gene Symbol:
Myl1
Gene Name:
myosin, light chain 1
Gene Aliases:
MLC3-f, MLCf, Mcl3, Mlc1, Mlc3
Chromosome Location:
Chr.9: 73937820 - 73958480 on Build Rnor_6.0
UniGene ID:
Rn.84920
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
191880
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Myl1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001077656.1
NP_001071124.1
5
612
NM_020104.2
NP_064489.1
5
430
XM_008767294.1
XP_008765516.1
4
560
XM_008767295.1
XP_008765517.1
4
681
GenBank mRNA
CR755322.1
-
5
435
CR756356.1
-
5
430
FQ214580.1
-
5
739
FQ214604.1
-
5
657
FQ214608.1
-
5
617
FQ214631.1
-
5
599
FQ214638.1
-
5
707
FQ214656.1
-
5
448
FQ214675.1
-
5
706
FQ214712.1
-
5
620
FQ214723.1
-
5
620
FQ214727.1
-
5
620
FQ214742.1
-
5
620
FQ214939.1
-
5
617
FQ215002.1
-
5
706
FQ215050.1
-
5
620
FQ215083.1
-
5
617
FQ215086.1
-
5
450
FQ215201.1
-
5
617
FQ215229.1
-
5
711
FQ215325.1
-
5
706
FQ215340.1
-
5
448
FQ215418.1
-
5
706
FQ215435.1
-
5
617
FQ215500.1
-
4
1196
FQ215535.1
-
4
681
FQ215557.1
-
5
706
FQ215563.1
-
5
614
FQ215578.1
-
5
706
FQ215610.1
-
5
620
FQ215748.1
-
4
679
FQ215766.1
-
5
706
FQ215774.1
-
5
617
FQ215820.1
-
5
675
FQ215848.1
-
5
617
FQ215875.1
-
5
617
FQ215911.1
-
5
711
FQ216034.1
-
5
620
FQ216060.1
-
5
706
FQ216109.1
-
5
617
FQ216124.1
-
5
711
FQ216181.1
-
5
599
FQ216192.1
-
5
706
FQ216218.1
-
5
617
FQ216229.1
-
5
887
FQ216238.1
-
5
706
FQ216407.1
-
5
788
FQ216452.1
-
5
617
FQ216455.1
-
5
621
FQ216484.1
-
5
599
FQ216496.1
-
5
620
FQ216525.1
-
5
620
FQ216543.1
-
5
617
FQ216556.1
-
5
706
FQ216567.1
-
5
617
FQ216631.1
-
5
620
FQ216654.1
-
5
704
FQ216704.1
-
5
620
FQ216737.1
-
5
703
FQ216738.1
-
5
448
FQ217074.1
-
5
447
FQ217103.1
-
5
448
FQ217157.1
-
4
423
FQ217192.1
-
5
448
FQ217218.1
-
5
448
FQ217243.1
-
5
447
FQ217303.1
-
5
599
FQ217323.1
-
5
448
FQ217387.1
-
5
448
FQ217394.1
-
5
550
FQ217427.1
-
5
453
FQ217481.1
-
5
448
FQ217524.1
-
5
448
FQ217753.1
-
5
453
FQ217756.1
-
5
448
FQ217889.1
-
5
448
FQ217973.1
-
5
453
FQ217979.1
-
5
448
FQ218029.1
-
5
448
FQ218049.1
-
5
445
FQ223641.1
-
5
706
FQ223687.1
-
5
448
FQ223755.1
-
5
616
FQ223779.1
-
5
448
FQ223827.1
-
5
620
FQ223880.1
-
5
518
FQ223949.1
-
5
446
FQ223984.1
-
5
448
FQ224047.1
-
5
448
FQ224050.1
-
5
448
FQ224087.1
-
5
448
FQ224112.1
-
4
423
FQ224155.1
-
5
453
FQ224171.1
-
5
617
FQ224172.1
-
5
448
FQ224212.1
-
5
600
FQ224254.1
-
5
448
FQ224277.1
-
5
620
FQ224321.1
-
5
452
FQ224386.1
-
5
453
FQ224411.1
-
5
448
FQ224420.1
-
5
448
FQ224452.1
-
5
523
FQ224519.1
-
5
617
FQ224645.1
-
2
48
FQ224663.1
-
5
448
FQ224726.1
-
5
550
FQ224796.1
-
5
448
FQ224799.1
-
5
453
FQ224867.1
-
5
706
FQ224927.1
-
5
706
FQ224931.1
-
5
617
FQ224934.1
-
5
615
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
cardiac muscle contraction
calcium ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-9ljwx:80/10.239.166.46:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0