Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020542.2 | NP_065417.1 | 2 | 970 | ||
GenBank mRNA | BC079207.1 | - | 2 | 970 | 7131790 |