Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mark2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021699.1 | NP_067731.1 | 2 | 124 | ||
XM_006230879.2 | XP_006230941.1 | 2 | 624 | |||
XM_006230880.2 | XP_006230942.1 | 2 | 1411 | |||
XM_006230881.2 | XP_006230943.1 | 2 | 143 | |||
XM_006230883.2 | XP_006230945.1 | 2 | 623 | |||
XM_006230884.2 | XP_006230946.1 | 2 | 622 | |||
XM_006230885.2 | XP_006230947.1 | 2 | 622 | |||
XM_006230887.2 | XP_006230949.1 | 2 | 621 | |||
XM_008760169.1 | XP_008758391.1 | 2 | 623 | |||
GenBank mRNA | Z83869.1 | - | 2 | 124 |