Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acrv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021747.1 | NP_068515.1 | 2 | 254 | ||
GenBank mRNA | AJ243484.1 | - | 2 | 254 |