Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spef2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022620.1 | NP_072142.1 | 4, 5 | 630 | ||
XM_008760793.1 | XP_008759015.1 | 5, 6 | 873 | |||
GenBank mRNA | AF102129.1 | - | 4, 5 | 630 | ||
BC097318.1 | - | 5, 6 | 785 |