Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp238 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022678.1 | NP_073169.1 | 1 | 176 | ||
XM_006250328.2 | XP_006250390.1 | 3 | 1269 | |||
XM_006250330.2 | XP_006250392.1 | 2 | 1185 | |||
XM_006250331.2 | XP_006250393.1 | 2 | 664 | |||
XM_008769796.1 | XP_008768018.1 | 2 | 1916 | |||
XM_008769797.1 | XP_008768019.1 | 3 | 1048 | |||
XM_008769798.1 | XP_008768020.1 | 3 | 1268 | |||
XM_008769799.1 | XP_008768021.1 | 2 | 3187 | |||
XM_008769800.1 | XP_008768022.1 | 2 | 219 | |||
GenBank mRNA | AF221838.1 | - | 1 | 176 |