Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022680.1 | NP_073171.1 | 2 | 89 |
Gene Symbol: Alppl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767355.1 | XP_008765577.1 | 3 | 1923 |