Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taok2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022702.1 | NP_073193.1 | 4 | 446 | ||
XM_006230329.2 | XP_006230391.1 | 4 | 1869 | |||
XM_006230330.2 | XP_006230392.1 | 3 | 290 | |||
XM_006230331.2 | XP_006230393.1 | 4 | 1157 | |||
XM_006230332.2 | XP_006230394.1 | 4 | 1158 | |||
XM_006230334.2 | XP_006230396.1 | 4 | 1142 | |||
GenBank mRNA | AB290408.1 | - | 3 | 254 | ||
AF140556.2 | - | 4 | 446 | |||
FQ223093.1 | - | 4 | 1065 |