Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fabp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022854.1 | NP_074045.1 | 4 | 415 | ||
GenBank mRNA | BC078817.1 | - | 4 | 416 | 7133719 | |
U07870.1 | - | 4 | 401 | |||
U09022.1 | - | 4 | 415 |