Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007058.3 | NP_008989.2 | 14 | 1507 | ||
XM_006714984.1 | XP_006715047.1 | 14 | 1510 | |||
XM_006714985.1 | XP_006715048.1 | 14 | 1510 | |||
XM_006714987.1 | XP_006715050.1 | 14 | 1510 | |||
XM_011514274.1 | XP_011512576.1 | 14 | 1591 | |||
XM_011514275.1 | XP_011512577.1 | 15 | 1633 | |||
GenBank mRNA | AJ242832.1 | - | 13 | 1461 | ||
AK300107.1 | - | 5 | 433 | |||
AK309883.1 | - | 4 | 819 | |||
AK314054.1 | - | 14 | 1487 | |||
BC033733.1 | - | 14 | 1478 | 5170266 |