Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lphn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022962.1 | NP_075251.1 | 5, 6 | 692 | ||
GenBank mRNA | AF081146.1 | - | 5, 6 | 692 | ||
AF081147.1 | - | 5, 6 | 692 | |||
AF111099.1 | - | 6, 7 | 1018 | |||
U72487.1 | - | 4, 5 | 494 |
Set Membership: |
G-protein coupled receptor |