Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030834.1 | NP_110461.1 | 3 | 426 | ||
XM_006247931.2 | XP_006247993.1 | 4 | 541 | |||
XM_006247932.2 | XP_006247994.1 | 4 | 833 | |||
XM_006247933.2 | XP_006247995.1 | 3 | 448 | |||
XM_006247934.2 | XP_006247996.1 | 4 | 507 | |||
XM_008768513.1 | XP_008766735.1 | 4 | 3265 | |||
XM_008768514.1 | XP_008766736.1 | 3 | 3172 | |||
GenBank mRNA | BC168146.1 | - | 3 | 385 | 9089640 | |
U87627.1 | - | 3 | 426 |