Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bdkrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030851.1 | NP_110478.1 | 2 | 1274 | ||
XM_008764861.1 | XP_008763083.1 | 1 | 1152 | |||
GenBank mRNA | AJ132230.2 | - | 2 | 1274 |
Set Membership: |
![]() |