Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007083.4 | NP_009014.2 | 2, 3 | 484 | ||
NM_198041.2 | NP_932158.1 | 2, 3 | 416 | |||
GenBank mRNA | AB209758.1 | - | 1, 2 | 5103 | ||
AF019632.2 | - | 2, 3 | 380 | |||
AF019633.2 | - | 2, 3 | 484 | |||
AK291871.1 | - | 2, 3 | 483 | |||
AK294450.1 | - | 2, 3 | 416 | |||
BC009842.2 | - | 2, 3 | 464 | 3942416 | ||
BG386740.1 | - | 2, 3 | 462 | |||
CB128685.1 | - | 2, 3 | 485 |