Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6ap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031785.1 | NP_113973.1 | 9 | 1082 | ||
GenBank mRNA | AF035387.1 | - | 9 | 1082 | ||
BC070937.1 | - | 9 | 1082 | 7099117 |