Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inhbe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031815.2 | NP_114003.2 | 1 | 289 | ||
GenBank mRNA | AF089825.1 | - | 480 | |||
AF140032.1 | - | 1 | 289 |