Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053541.1 | NP_445993.1 | 3 | 594 | ||
XM_008759197.1 | XP_008757419.1 | 2 | 1754 | |||
XM_008759198.1 | XP_008757420.1 | 2 | 1754 | |||
XM_008759199.1 | XP_008757421.1 | 2 | 1754 | |||
XM_008759200.1 | XP_008757422.1 | 3 | 673 | |||
XM_008759201.1 | XP_008757423.1 | 2 | 1753 | |||
XM_008759202.1 | XP_008757424.1 | 3 | 673 | |||
GenBank mRNA | AB009463.1 | - | 3 | 594 |