Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1b7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053781.2 | NP_446233.2 | 10 | 939 | ||
XM_006236250.2 | XP_006236312.1 | 11 | 1289 | |||
XM_008762755.1 | XP_008760977.1 | 9 | 1794 | |||
GenBank mRNA | AF182168.1 | - | 10 | 1081 | ||
BC086563.1 | - | 10 | 939 | 7307075 |