Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F11r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053796.1 | NP_446248.1 | 3, 4 | 308 | ||
GenBank mRNA | AF276998.1 | - | 3, 4 | 308 | ||
BC065309.1 | - | 3, 4 | 290 | 5599898 |