Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brinp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080482.2 | NP_536730.1 | 1 | 157 | ||
XM_006238272.2 | XP_006238334.1 | 2 | 575 | |||
GenBank mRNA | AB051356.1 | - | 1 | 158 |