Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SYNRG SIRNA Assays ›
Gene Symbol:
SYNRG
Gene Name:
synergin, gamma
Gene Aliases:
AP1GBP1, SYNG
Chromosome Location:
Chr.17: 37514797 - 37613637 on Build GRCh38
UniGene ID:
Hs.594647
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
19327
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SYNRG
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001163544.2
NP_001157016.1
8
852
NM_001163545.2
NP_001157017.1
8
849
NM_001163546.2
NP_001157018.1
8
852
NM_001163547.2
NP_001157019.1
8
852
NM_007247.5
NP_009178.3
9
1086
NM_080550.4
NP_542117.3
8
852
NM_198882.2
NP_942583.1
8
852
XM_005256980.4
XP_005257037.1
10
1447
XM_017024076.1
XP_016879565.1
10
1447
XM_017024077.1
XP_016879566.1
10
1366
XM_017024078.1
XP_016879567.1
10
1447
XM_017024079.1
XP_016879568.1
10
1447
XM_017024080.1
XP_016879569.1
10
5499
XM_017024081.1
XP_016879570.1
10
5496
XM_017024082.1
XP_016879571.1
10
1447
XM_017024083.1
XP_016879572.1
10
1447
XM_017024084.1
XP_016879573.1
10
1447
XM_017024085.1
XP_016879574.1
10
1447
XM_017024086.1
XP_016879575.1
10
1447
XM_017024087.1
XP_016879576.1
10
1447
XM_017024088.1
XP_016879577.1
9
1213
XM_017024089.1
XP_016879578.1
10
1447
XM_017024090.1
XP_016879579.1
9
1144
XM_017024091.1
XP_016879580.1
9
1063
XM_017024092.1
XP_016879581.1
10
1447
XM_017024093.1
XP_016879582.1
9
1144
XM_017024094.1
XP_016879583.1
9
1063
XM_017024095.1
XP_016879584.1
9
1144
XM_017024096.1
XP_016879585.1
10
1447
XM_017024097.1
XP_016879586.1
9
1144
XM_017024098.1
XP_016879587.1
9
1144
XM_017024099.1
XP_016879588.1
8
829
XM_017024100.1
XP_016879589.1
9
1144
XM_017024101.1
XP_016879590.1
9
1063
XM_017024102.1
XP_016879591.1
8
829
XM_017024103.1
XP_016879592.1
8
910
XM_017024104.1
XP_016879593.1
8
910
XM_017024105.1
XP_016879594.1
10
1447
XM_017024106.1
XP_016879595.1
8
910
GenBank mRNA
AF169548.1
-
9
1046
AK025335.1
-
2
242
AK126988.1
-
9
1814
AK302821.1
-
10
1324
AK308292.1
-
3
465
BC090930.1
-
8
803
3077104
BC117313.1
-
8
792
40125864
BC143476.1
-
8
795
9051984
BC143478.1
-
8
795
9051986
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_053419, EF121981, EF121979, EF121980, EF121978, and others.
Gene Ontology Categories:
Function(s)
Process(es)
intracellular protein transport
endocytosis
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-j4kkz:80/10.239.163.216:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0