Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd79b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133533.2 | NP_598217.1 | 6 | 680 | ||
GenBank mRNA | BC088115.1 | - | 6 | 680 | 7325406 | |
FQ226266.1 | - | 6 | 677 | |||
FQ227061.1 | - | 6 | 675 | |||
FQ227788.1 | - | 6 | 672 | |||
FQ230261.1 | - | 6 | 671 | |||
FQ232054.1 | - | 6 | 671 | |||
FQ232680.1 | - | 6 | 671 | |||
FQ232790.1 | - | 6 | 694 | |||
FQ233507.1 | - | 6 | 669 | |||
FQ234082.1 | - | 6 | 654 | |||
FQ234487.1 | - | 6 | 669 |