Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcsk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133559.1 | NP_598243.1 | 6 | 630 | ||
XM_006240839.2 | XP_006240901.1 | 6 | 816 | |||
XM_006240843.2 | XP_006240905.1 | 6 | 816 | |||
XM_008765083.1 | XP_008763305.1 | 6 | 816 | |||
XM_008765084.1 | XP_008763306.1 | 6 | 816 | |||
XM_008765085.1 | XP_008763307.1 | 6 | 816 | |||
XM_008765086.1 | XP_008763308.1 | 6 | 783 | |||
XM_008765087.1 | XP_008763309.1 | 6 | 816 | |||
XM_008765088.1 | XP_008763310.1 | 6 | 816 | |||
GenBank mRNA | BC097288.1 | - | 6 | 642 | 7456182 | |
L14937.1 | - | 6 | 630 |