Search Thermo Fisher Scientific
Gene Symbol: Slc2a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133611.2 | NP_598295.2 | 5 | 1172 | ||
XM_008765664.1 | XP_008763886.1 | 2 | 236 | |||
GenBank mRNA | AJ315643.1 | - | 5 | 1180 |