Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUSC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007275.2 | NP_009206.1 | 2 | 342 | ||
GenBank mRNA | AF055479.1 | - | 2 | 350 | ||
AK124058.1 | - | 3 | 276 | |||
AK311998.1 | - | 2 | 332 | |||
BC023976.2 | - | 2 | 294 | 3504783 | ||
CA392689.1 | - | 2 | 342 |