Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100910790 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770410.1 | XP_008768632.1 | 454 |
Gene Symbol: Spon2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138533.3 | NP_612542.1 | 2 | 338 | ||
GenBank mRNA | BC078734.1 | - | 2 | 338 | 7121065 |