Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tecr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138549.1 | NP_612558.1 | 3 | 248 | ||
XM_006255216.2 | XP_006255278.1 | 2 | 383 | |||
XM_006255217.2 | XP_006255279.1 | 2 | 295 | |||
XM_006255218.2 | XP_006255280.1 | 3 | 253 | |||
GenBank mRNA | BC059115.1 | - | 3 | 211 | 6920522 | |
EU000473.1 | - | 3 | 211 | |||
FQ212009.1 | - | 3 | 234 | |||
FQ215654.1 | - | 2 | 305 | |||
FQ215778.1 | - | 1 | 115 | |||
FQ216428.1 | - | 2 | 300 | |||
FQ219100.1 | - | 3 | 188 | |||
FQ220726.1 | - | 3 | 197 | |||
S45663.1 | - | 3 | 248 |