Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138881.1 | NP_620236.1 | 1 | 249 | ||
GenBank mRNA | Y07704.1 | - | 1 | 249 |